Invitrogen
Manufacturer Code:PA586771
Catalog # PIPA586771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human PI3K p85beta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ca++/calmodulin-dependent protein kinase II beta subunit; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase type II beta chain; Calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; CaM kinase II subunit beta; CaM-kinase II beta chain; CaMK II; caMK-II subunit beta; Phosphatidylinositol 3-kinase regulatory subunit beta PI3-kinase regulatory subunit beta PI3K regulatory subunit beta PtdIns-3-kinase regulatory subunit beta Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta PI3-kinase subunit p85-beta PtdIns-3-kinase regulatory subunit p85-beta PIK3R2; proline rich calmodulin-dependent protein kinase
Gene Aliases: CAM2; CAMK2; CAMK2B; Camk2d; CAMKB; Ck2b
UniProt ID: (Human) Q13554, (Rat) P08413, (Mouse) P28652
Entrez Gene ID: (Human) 816, (Rat) 24245, (Mouse) 12323
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.