Novus Biologicals
Manufacturer Code:NBP215684
Catalog # NBP215684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CAMK2B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase (CaM kinase) II beta calcium/calmodulin-dependent protein kinase II beta calcium/calmodulin-dependent protein kinase type II beta chain calcium/calmodulin-dependent protein kinase type II subunit beta CaM kinase II beta subunit CaM kinase II subunit beta CAM2CAMK2 CAMKB CaMK-II subunit beta CaM-kinase II beta chain EC 2.7.11 EC 2.7.11.17 MGC29528 proline rich calmodulin-dependent protein kinase; calcium/calmodulin-dependent protein kinase II beta; Calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; CaM kinase II subunit beta; CaM-kinase II beta chain; caMK-II subunit beta; proline rich calmodulin-dependent protein kinase
Gene Aliases: CAM2; CAMK2; CAMK2B; CAMKB
UniProt ID: (Human) Q13554
Entrez Gene ID: (Human) 816
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.