Invitrogen
Manufacturer Code:PA517095
Catalog # PIPA517095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the human sequence of CaMKII-a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; Calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; CaM kinase II alpha subunit CaM-kinase II alpha chain CaMK-II alpha subunit CaMKIINalpha caM kinase II subunit alpha CaMK-II subunit alpha calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha calcium/calmodulin-dependent protein kinase II alpha-B subunit calcium/calmodulin-dependent protein kinase type II alpha chain calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II subunit alpha; CaM-kinase II alpha chain; CaMK II; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha
Gene Aliases: CAMK2A; CAMKA; CaMKII; KIAA0968; mKIAA0968; PK2CDD; PKCCD; R74975
UniProt ID: (Human) Q9UQM7, (Mouse) P11798, (Rat) P11275
Entrez Gene ID: (Human) 815, (Mouse) 12322, (Rat) 25400
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.