Invitrogen
Manufacturer Code:PA514032
Catalog # PIPA514032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 341-370 amino acids from the C-terminal region of human CAMK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase I; Calcium/calmodulin-dependent protein kinase type 1; CAM kinase I; CaM kinase I alpha; CaM-KI; CaMKI-alpha; G alpha interacting protein; regulator of G-protein signalling 19
Gene Aliases: AI505105; CAMK1; CAMKI; CaMKIalpha; D6Ertd263e; Gaip
UniProt ID: (Human) Q14012, (Rat) Q63450, (Mouse) Q91YS8
Entrez Gene ID: (Human) 8536, (Rat) 171503, (Mouse) 52163
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.