Invitrogen
Manufacturer Code:PA522766
Catalog # PIPA522766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCN family member 1; CCN1 GIG1 IGFBP10 CCN family member 1 IBP-10 IGF-binding protein 10 IGFBP-10 cysteine-rich heparin-binding protein 61 cysteine-rich anigogenic inducer 61 insulin-like growth factor-binding protein 10 protein CYR61; Cellular communication network factor 1; cysteine rich angiogenic inducer 61; Cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; IBP-10; IGF-binding protein 10; IGFBP-10; Insulin-like growth factor-binding protein 10; Protein CYR61; Protein GIG1
Gene Aliases: CCN1; CYR61; GIG1; IGFBP10
UniProt ID: (Human) O00622
Entrez Gene ID: (Rabbit) 100339657, (Human) 3491
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.