Invitrogen
Manufacturer Code:PA5114440
Catalog # PIPA5114440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CCN1 recombinant protein (Position: K54-D381). Human CCN1 shares 91.8% and 92.1% amino acid (aa) sequence identity with mouse and rat CCN1 respectively. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3CH61 AI325051 CCN family member 1 Ccn1 Cellular communication network factor 1 Cyr61 cysteine rich angiogenic inducer 61 cysteine rich protein 61 cysteine rich protein 61-like cysteine-rich angiogenic inducer 61 cysteine-rich heparin-binding protein 61 cysteine-rich angiogenic inducer 61 cysteine-rich anigogenic inducer 61 GIG1 IBP-10 IGF-binding protein 10 IGFBP10 IGFBP-10 insulin-like growth factor binding protein 10 insulin-like growth factor-binding protein 10 intermediate early LOC100339657 protein CYR61 Protein GIG1; CCN family member 1; Cellular communication network factor 1; cysteine rich protein 61; Cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; IBP-10; IGF-binding protein 10; IGFBP-10; Insulin-like growth factor-binding protein 10; Protein CYR61; Protein GIG1
Gene Aliases: CCN1; CYR61; GIG1; IGFBP10
UniProt ID: (Human) O00622, (Rat) Q9ES72
Entrez Gene ID: (Human) 3491, (Rat) 83476
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.