Novus Biologicals
Manufacturer Code:NBP250217AF594
Catalog # NP20217A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M17-P3F2 |
Immunogen | Ovalbumin-conjugated synthetic peptide IQYPDSDVL (C-terminal sequence) |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 24-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; CBAS3 CP7B Cytochrome P450 7B1 cytochrome P450 family 7 subfamily B polypeptide 1 cytochrome P450 subfamily VIIB (oxysterol 7 alpha-hydroxylase) polypeptide 125-hydroxycholesterol 7-alpha-hydroxylase EC 1.14.13.100 oxysterol 7alpha-hydroxylase Oxysterol 7-alpha-hydroxylase spastic paraplegia 5A (autosomal recessive) SPG5A; Cytochrome P450 7B1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; Oxysterol 7-alpha-hydroxylase
Gene Aliases: CBAS3; CP7B; CYP7B1; SPG5A
UniProt ID: (Human) O75881
Entrez Gene ID: (Human) 9420
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.