Proteintech
Manufacturer Code:248891AP150UL
Catalog # 50-173-4598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP7B1 Fusion Protein Ag19374 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24-hydroxycholesterol 7-alpha-hydroxylase; 25-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; CBAS3 CP7B CYP7B1 Cytochrome P450 7B1 Oxysterol 7 alpha hydroxylase SPG5A; Cytochrome P450 7B1; cytochrome P450, 7b1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; HCT-1; Hippocampal transcript 1 protein; Oxysterol 7-alpha-hydroxylase
Gene Aliases: AW261589; CBAS3; CP7B; CYP7B1; D3Ertd552e; hct-1; SPG5A
UniProt ID: (Human) O75881, (Mouse) Q60991
Entrez Gene ID: (Human) 9420, (Mouse) 13123
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.