Invitrogen
Manufacturer Code:PA579135
Catalog # PIPA579135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CYP7A1 recombit protein (Position: M301-L504). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24-hydroxycholesterol 7-alpha-hydroxylase; cholesterol 7 alpha hydroxylase; cholesterol 7-alpha hydroxylase; Cholesterol 7-alpha-hydroxylase; Cholesterol 7-alpha-monooxygenase; CYPVII; cytochrome P450 (cholesterol hydroxylase 7 alpha); Cytochrome P450 7A1; cytochrome P450, 7a1; cytochrome P450, family 7, subfamily A, polypeptide 1; cytochrome P450, subfamily VIIA polypeptide 1
Gene Aliases: CHAP; CP7A; CYP7; CYP7A1; CYP7S1; CYPVII
UniProt ID: (Human) P22680, (Mouse) Q64505, (Rat) P18125
Entrez Gene ID: (Human) 1581, (Mouse) 13122, (Rat) 25428
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.