Novus Biologicals
Manufacturer Code:NBP30483620UL
Catalog # NB094495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-280 of human CYP7A1 (NP_000771.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24-hydroxycholesterol 7-alpha-hydroxylase; Cholesterol 7-alpha-hydroxylase; Cholesterol 7-alpha-hydroxylase cholesterol 7-alpha-monooxygenase CP7A CYP7MGC126826 CYPVII Cytochrome P450 7A1 cytochrome P450 family 7 subfamily A polypeptide 1 cytochrome P450 subfamily VIIA (cholesterol 7 alpha-monooxygenase) cytochrome P450 subfamily VIIA polypeptide 1 EC 1.14.13 EC 1.14.13.17 MGC138389 polypeptide 1; Cholesterol 7-alpha-monooxygenase; CYPVII; Cytochrome P450 7A1; cytochrome P450, family 7, subfamily A, polypeptide 1; cytochrome P450, subfamily VIIA polypeptide 1
Gene Aliases: CP7A; CYP7; CYP7A1; CYPVII
UniProt ID: (Human) P22680
Entrez Gene ID: (Human) 1581
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.