Invitrogen
Manufacturer Code:PA576873
Catalog # PIPA576873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CYP51A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CP51CP51A&lowbarHUMANCYP51CYP51A1CYPL1CYPLICytochrome P450 51Cytochrome P450 51A1Cytochrome P450 family 51 subfamily A polypeptide 1Cytochrome P450-14DMCytochrome P45014DMCytochrome P450LILanosterol 14-alpha demethylaseLDMP450 14DMP450L1Sterol 14-alpha demethylase; CYPLI; Cytochrom P450 Lanosterol 14 alpha-demethylase; Cytochrome P450 51A1; cytochrome P450 Lanosterol 14 alpha-demethylase; cytochrome P450, 51; cytochrome P450, 51 (lanosterol 14-alpha-demethylase); cytochrome P450, family 51, subfamily A, polypeptide 1; cytochrome P450, subfamily 51; Cytochrome P450-14DM; Cytochrome P45014DM; Cytochrome P450LI; Lanosterol 14-alpha demethylase; lanosterol 14-alpha-demethylase; LDM; P450-14DM; P45014DM; P450LI; Sterol 14-alpha demethylase
Gene Aliases: AI426508; CP51; CYP51; CYP51A1; CYPL1; CYPLI; LDM; P450-14DM; P450L1; P450LI; RATCP14DM
UniProt ID: (Human) Q16850, (Mouse) Q8K0C4, (Rat) Q64654
Entrez Gene ID: (Human) 1595, (Mouse) 13121, (Rat) 25427
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.