Proteintech
Manufacturer Code:134311AP150UL
Catalog # 50-172-8739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP51A1 Fusion Protein Ag4235 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CP51 CYP51 CYP51A1 CYPL1 CYPLI Cytochrome P450 14DM Cytochrome P450 51A1 Cytochrome P45014DM Cytochrome P450LI LDM P450 14DM P450L1 Sterol 14 alpha demethylase; CYPLI; Cytochrom P450 Lanosterol 14 alpha-demethylase; Cytochrome P450 51A1; cytochrome P450 Lanosterol 14 alpha-demethylase; cytochrome P450, 51; cytochrome P450, 51 (lanosterol 14-alpha-demethylase); cytochrome P450, family 51, subfamily A, polypeptide 1; cytochrome P450, subfamily 51; Cytochrome P450-14DM; Cytochrome P45014DM; Cytochrome P450LI; Lanosterol 14-alpha demethylase; lanosterol 14-alpha-demethylase; LDM; P450-14DM; P45014DM; P450LI; Sterol 14-alpha demethylase
Gene Aliases: AI426508; CP51; CYP51; CYP51A1; CYPL1; CYPLI; LDM; P450-14DM; P450L1; P450LI; RATCP14DM
UniProt ID: (Human) Q16850, (Mouse) Q8K0C4, (Rat) Q64654
Entrez Gene ID: (Human) 1595, (Mouse) 13121, (Rat) 25427
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.