Abnova
Manufacturer Code:H00011283K
Catalog # 89-201-083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CYP4F8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CYP4F8 peptide using ARM Technology.
For Research Use Only
Protein Aliases: CPF8 CYPIVF8; CYPIVF8; Cytochrome P450 4F8; cytochrome P450, family 4, subfamily F, polypeptide 8; cytochrome P450, subfamily IVF, polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase
Gene Aliases: CPF8; CYP4F8; CYPIVF8
UniProt ID: (Human) P98187
Entrez Gene ID: (Human) 11283
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.