Abnova
Manufacturer Code:PAB14598
Catalog # 89-117-173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 354-371 of human CYP4F3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Protein Aliases: 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; CYPIVF3; cytochrome P-450; Cytochrome P450 4F3; cytochrome P450, family 4, subfamily F, polypeptide 3; cytochrome P450, subfamily IVF, polypeptide 3 (leukotriene B4 omega hydroxylase); Cytochrome P450-LTB-omega; Docosahexaenoic acid omega-hydroxylase CYP4F3; leukotriene B4 omega hydroxylase; Leukotriene-B(4) 20-monooxygenase 2; Leukotriene-B(4) omega-hydroxylase 2; leukotriene-B4 20-monooxygenase
Gene Aliases: CPF3; CYP4F; CYP4F3; LTB4H
UniProt ID: (Human) Q08477
Entrez Gene ID: (Human) 4051
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.