Proteintech
Manufacturer Code:197011AP150UL
Catalog # 50-173-2498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; CPF3 CYP4F CYP4F3 CYPIVF3 Cytochrome P450 4F3 Cytochrome P450 LTB omega LTB4H; CYPIVF3; cytochrome P-450; Cytochrome P450 4F3; cytochrome P450, family 4, subfamily F, polypeptide 3; cytochrome P450, subfamily IVF, polypeptide 3 (leukotriene B4 omega hydroxylase); Cytochrome P450-LTB-omega; Docosahexaenoic acid omega-hydroxylase CYP4F3; leukotriene B4 omega hydroxylase; Leukotriene-B(4) 20-monooxygenase 2; Leukotriene-B(4) omega-hydroxylase 2; leukotriene-B4 20-monooxygenase
Gene Aliases: CPF3; CYP4F; CYP4F3; LTB4H
UniProt ID: (Human) Q08477
Entrez Gene ID: (Human) 4051
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.