Invitrogen
Manufacturer Code:PA599614
Catalog # PIPA599614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CYP4F2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20-HETE synthase; 20-HETE synthase 20-hydroxyeicosatetraenoic acid synthase Arachidonic acid omega-hydroxylase CPF2 CYP4F2 CYPIVF2 Cytochrome P450 4F2 cytochrome P450 family 4 subfamily F member 2 cytochrome P450 family 4 subfamily F polypeptide 2 cytochrome P450 subfamily IVF polypeptide 2 cytochrome P450-LTB-omega Docosahexaenoic acid omega-hydroxylase docosahexaenoic acid omega-hydroxylase CYP4F3 phylloquinone omega-hydroxylase CYP4F2 leukotriene-B(4) 20-monooxygenase 1 Leukotriene-B(4) omega-hydroxylase 1 phylloquinone omega-hydroxylase CYP4F2; 20-hydroxyeicosatetraenoic acid synthase; Arachidonic acid omega-hydroxylase; CYPIVF2; Cytochrome P450 4F2; cytochrome P450, family 4, subfamily F, polypeptide 2; cytochrome P450, subfamily IVF, polypeptide 2; Cytochrome P450-LTB-omega; Docosahexaenoic acid omega-hydroxylase; Leukotriene-B(4) 20-monooxygenase 1; Leukotriene-B(4) omega-hydroxylase 1; Phylloquinone omega-hydroxylase CYP4F2
Gene Aliases: CPF2; CYP4F2
UniProt ID: (Human) P78329
Entrez Gene ID: (Human) 8529
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.