Proteintech
Manufacturer Code:132431AP150UL
Catalog # 50-172-8598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP4F12 Fusion Protein Ag3957 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYP4F12 CYPIVF12 Cytochrome P450 4F12 F22329&lowbar1; CYPIVF12; Cytochrome P450 4F12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12
Gene Aliases: CYP4F12; CYPIVF12; F22329_1; UNQ568/PRO1129
UniProt ID: (Human) Q9HCS2
Entrez Gene ID: (Human) 66002
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.