Invitrogen
Manufacturer Code:MA112890
Catalog # 01-674-091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F21 P6 F5 |
Immunogen | Synthetic peptide corresponding to human CYP450 4F11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy fatty acids omega-hydroxylase CYP4F11; CYPIVF11; Cytochrome P450 4F11; cytochrome P450, family 4, subfamily F, polypeptide 11; cytochrome P450, subfamily IVF, polypeptide 11; Docosahexaenoic acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; Phylloquinone omega-hydroxylase CYP4F11
Gene Aliases: CYP4F11; CYPIVF11
UniProt ID: (Human) Q9HBI6
Entrez Gene ID: (Human) 57834
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.