Invitrogen
Manufacturer Code:PA5113387
Catalog # PIPA5113387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phylloquinone omega-hydroxylase CYP4F11 protein (38-524AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy fatty acids omega-hydroxylase CYP4F11; 3-hydroxy fatty acids omega-hydroxylase CYP4F11 CYP4F11 CYPIVF11 Cytochrome P450 4F11 cytochrome P450 family 4 subfamily F member 11 cytochrome P450 family 4 subfamily F polypeptide 11 cytochrome P450 subfamily IVF polypeptide 11 Docosahexaenoic acid omega-hydroxylase Long-chain fatty acid omega-monooxygenase Phylloquinone omega-hydroxylase CYP4F11; CYPIVF11; Cytochrome P450 4F11; cytochrome P450, family 4, subfamily F, polypeptide 11; cytochrome P450, subfamily IVF, polypeptide 11; Docosahexaenoic acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; Phylloquinone omega-hydroxylase CYP4F11
Gene Aliases: CYP4F11; CYPIVF11
UniProt ID: (Human) Q9HBI6
Entrez Gene ID: (Human) 57834
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.