Invitrogen
Manufacturer Code:PA569151
Catalog # PIPA569151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse CYP4B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYPIVB1; CYPIVB1 P-450HP cytochrome P450 4B1 cytochrome P450 subfamily IVB polypeptide 1 cytochrome P450-HP microsomal monooxygenase; cytochrome P-450 isozyme 5; Cytochrome P450 4B1; Cytochrome P450 isozyme 5; Cytochrome P450 L-2; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily 4B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; Cytochrome P450-HP; microsomal monooxygenase; P450 L-2; P450-isozyme 5
Gene Aliases: Cyp4b-1; CYP4B1; CYPIVB1; P-450HP
UniProt ID: (Rabbit) P15128, (Human) P13584, (Mouse) Q64462, (Rat) P15129
Entrez Gene ID: (Rabbit) 100008805, (Dog) 608452, (Human) 1580, (Bovine) 540149, (Mouse) 13120, (Rat) 24307
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.