Invitrogen
Manufacturer Code:PA5106560
Catalog # PIPA5106560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Cytochrome P450 4B1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyp4b1 Cyp4b-1 CYPIVB1 Cytochrome P450 4B1 cytochrome P450 family 4 subfamily B member 1 cytochrome P450 isozyme 5 Cytochrome P450 L-2 cytochrome P450 family 4 subfamily b polypeptide 1 cytochrome P450 subfamily 4B polypeptide 1 cytochrome P450 subfamily IV B polypeptide 1 cytochrome P450 subfamily IVB polypeptide 1 cytochrome P450-HP microsomal monooxygenase P450 L-2 P-450HP P450-isozyme 5; CYPIVB1; Cytochrome P450 4B1; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; Cytochrome P450-HP; microsomal monooxygenase
Gene Aliases: CYP4B1; CYPIVB1; P-450HP
UniProt ID: (Human) P13584, (Mouse) Q64462
Entrez Gene ID: (Human) 1580, (Mouse) 13120
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.