Proteintech
Manufacturer Code:117711AP150UL
Catalog # 50-172-7441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP4B1 Fusion Protein Ag2416 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYP4B1 CYPIVB1 Cytochrome P450 4B1 Cytochrome P450 HP P 450HP; CYPIVB1; Cytochrome P450 4B1; Cytochrome P450 isozyme 5; Cytochrome P450 L-2; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily 4B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; Cytochrome P450-HP; microsomal monooxygenase; P450 L-2; P450-isozyme 5
Gene Aliases: Cyp4b-1; CYP4B1; CYPIVB1; P-450HP
UniProt ID: (Human) P13584, (Mouse) Q64462, (Rat) P15129
Entrez Gene ID: (Human) 1580, (Mouse) 13120, (Rat) 24307
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.