Invitrogen
Manufacturer Code:MA537946
Catalog # PIMA537946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2219 |
Immunogen | A synthesized peptide derived from human Cytochrome P450 4A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 20-HETE synthase; 20-HETE synthase 20-hydroxyeicosatetraenoic acid synthase alkane-1 monooxygenase CP4Y CYP4A11 Cyp4a2 Cyp4a-2 CYP4AII CYPIVA11 CYPIVA2 Cytochrome P450 4A11 cytochrome P450 4A2 cytochrome P450 family 4 subfamily A member 11 Cytochrome P-450 K-2 cytochrome P450 K-5 cytochrome P450 family 4 subfamily A polypeptide 11 cytochrome P450 family 4 subfamily a polypeptide 2 cytochrome P450 subfamily 4A polypeptide 11 cytochrome P450 subfamily IVA polypeptide 11 cytochrome P-450HK-omega Cytochrome P450HL-omega cytochrome P450-LA-omega 2 fatty acid omega-hydroxylase Lauric acid omega-hydroxylase long-chain fatty acid omega-monooxygenase P-450 HK omega P-450 IV4AII P-450 K-2 P450 K-5 P450-HL-omega P450HL-omega P450-LA-omega 2; 20-hydroxyeicosatetraenoic acid synthase; alkane-1 monooxygenase; CYP4AII; CYPIVA11; CYPIVA2; Cytochrome P-450 K-2; Cytochrome P-450HK-omega; Cytochrome P450 4A11; Cytochrome P450 4A2; Cytochrome P450 K-5; cytochrome P450, family 4, subfamily A, polypeptide 11; cytochrome P450, subfamily 4A, polypeptide 11; cytochrome P450, subfamily IVA, polypeptide 11; Cytochrome P450-LA-omega 2; Cytochrome P450HL-omega; Fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; P-450 IV4AII; P-450 K-2; P450 K-5; P450-LA-omega 2; P450HL-omega
Gene Aliases: CP4Y; Cyp4a-2; CYP4A11; CYP4A2; CYP4AII
UniProt ID: (Human) Q02928, (Rat) P20816
Entrez Gene ID: (Human) 1579, (Rat) 24306
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.