Abnova
Manufacturer Code:H00001579K
Catalog # 89-208-041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CYP4A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CYP4A11 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; alkane-1 monooxygenase; CP4Y CYP4A2 CYP4AII; CYP4AII; CYPIVA11; Cytochrome P-450HK-omega; Cytochrome P450 4A11; cytochrome P450, family 4, subfamily A, polypeptide 11; cytochrome P450, subfamily IVA, polypeptide 11; Cytochrome P450HL-omega; Fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; P450HL-omega
Gene Aliases: CP4Y; CYP4A11; CYP4A2; CYP4AII
UniProt ID: (Human) Q02928
Entrez Gene ID: (Human) 1579
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.