Invitrogen
Manufacturer Code:MA3032
Catalog # 01-670-129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F24 P2 B10 |
Immunogen | Synthetic peptide corresponding to residues E S R D G T V S G A conjugated to ovalbumin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1,4-cineole 2-exo-monooxygenase; 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; aryl hydrocarbon hydroxylase; Cholesterol 25-hydroxylase; CYP3A4 CYP3A5 CP35 P450PCN3 PCN3; CYPIIIA28; CYPIIIA3; CYPIIIA4; CYPIIIA5; cytochrome P450 3A28; Cytochrome P450 3A3; Cytochrome P450 3A4; Cytochrome P450 3A5; Cytochrome P450 HLp; cytochrome P450 HLp2; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; Cytochrome P450-PCN1; Cytochrome P450-PCN3; flavoprotein-linked monooxygenase; glucocorticoid-inducible P450; microsomal monooxygenase; Nifedipine oxidase; P450-III, steroid inducible; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase; xenobiotic monooxygenase
Gene Aliases: CP33; CP34; CP35; CYP3A; CYP3A28; CYP3A3; CYP3A4; CYP3A5; CYPIIIA3; CYPIIIA4; CYPIIIA5; HLP; NF-25; P450C3; P450PCN1; P450PCN3; PCN3
UniProt ID: (Human) Q16757, (Human) P20815
Entrez Gene ID: (Human) 1576, (Bovine) 507988, (Bovine) 282214, (Human) 1577
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.