Invitrogen
Manufacturer Code:PA5101318
Catalog # PIPA5101318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Cytochrome P450 3A4/5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-cineole 2-exo-monooxygenase; 1,8-cineole 2-exo-monooxygenase; 18-cineole 2-exo-monooxygenase albendazole monooxygenase Albendazole monooxygenase (sulfoxide-forming) Albendazole sulfoxidase aryl hydrocarbon hydroxylase Cholesterol 25-hydroxylase CP33 CP34 CP35 CYP3A CYP3A28 CYP3A3 CYP3A4 CYP3A5 CYPIIIA28 CYPIIIA3 CYPIIIA4 CYPIIIA5 cytochrome P450 cytochrome P450 3A28 cytochrome P450 3A3 Cytochrome P450 3A4 cytochrome P450 3A5 cytochrome P450 family 3 subfamily A member 4 cytochrome P450 family 3 subfamily A member 5 cytochrome P450 HLp cytochrome P450 HLp2 cytochrome P450 NF-25 cytochrome P450 family 3 subfamily A polypeptide 4 cytochrome P450 family 3 subfamily A polypeptide 5 cytochrome P450 subfamily IIIA (niphedipine oxidase) polypeptide 3 cytochrome P450 subfamily IIIA (niphedipine oxidase) polypeptide 4 cytochrome P450 subfamily IIIA (niphedipine oxidase) polypeptide 5 cytochrome P450 subfamily IIIA polypeptide 4 cytochrome P450-PCN1 Cytochrome P450-PCN3 flavoprotein-linked monooxygenase glucocorticoid-inducible P450 HLP HLp2 microsomal monooxygenase NF-25 Nifedipine oxidase P450C3 P450-III steroid inducible P450PCN1 P450PCN3 P450-PCN3 PCN3 Quinine 3-monooxygenase taurochenodeoxycholate 6-alpha-hydroxylase Unknown (protein for MGC:127840) xenobiotic monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; aryl hydrocarbon hydroxylase; Cholesterol 25-hydroxylase; CYPIIIA3; CYPIIIA4; CYPIIIA5; Cytochrome P450 3A3; Cytochrome P450 3A4; Cytochrome P450 3A5; Cytochrome P450 HLp; cytochrome P450 HLp2; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; Cytochrome P450-PCN1; Cytochrome P450-PCN3; flavoprotein-linked monooxygenase; glucocorticoid-inducible P450; microsomal monooxygenase; Nifedipine oxidase; P450-III, steroid inducible; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase; xenobiotic monooxygenase
Gene Aliases: CP33; CP34; CP35; CYP3A; CYP3A3; CYP3A4; CYP3A5; CYPIIIA3; CYPIIIA4; CYPIIIA5; HLP; NF-25; P450C3; P450PCN1; P450PCN3; PCN3
UniProt ID: (Human) Q16757, (Human) P20815
Entrez Gene ID: (Human) 1576, (Human) 1577
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.