Abnova
Manufacturer Code:MAB22020
Catalog # 89-339-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ACDE-3 |
Immunogen | A synthetic peptide corresponding to human CYP3A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: 1,4-cineole 2-exo-monooxygenase; 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; Cholesterol 25-hydroxylase; CP33 CP34 CYP3A CYP3A3 HLP MGC126680 NF-25 P450C3 P450PCN1; CYPIIIA3; CYPIIIA4; Cytochrome P450 3A3; Cytochrome P450 3A4; Cytochrome P450 HLp; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; Cytochrome P450-PCN1; glucocorticoid-inducible P450; Nifedipine oxidase; P450-III, steroid inducible; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase
Gene Aliases: CP33; CP34; CYP3A; CYP3A3; CYP3A4; CYPIIIA3; CYPIIIA4; HLP; NF-25; P450C3; P450PCN1
UniProt ID: (Human) Q16757
Entrez Gene ID: (Human) 1576
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.