Novus Biologicals
Manufacturer Code:NBP2839870.1ML
Catalog # NB031916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle terminal region of mouse CYP39A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24-hydroxycholesterol 7-alpha-hydroxylase; Cytochrome P450 39A1; Cytochrome P450 39A1 cytochrome P450 family 39 subfamily A polypeptide 1 cytochrome P450 subfamily XXXIX (oxysterol 7 alpha-hydroxylase) polypeptide 124-hydroxycholesterol 7-alpha-hydroxylase EC 1.14.13.99 hCYP39A1 oxysterol 7alpha-hydroxylase Oxysterol 7-alpha-hydroxylase; cytochrome P450, family 39, subfamily A, polypeptide 1; cytochrome P450, subfamily XXXIX (oxysterol 7 alpha-hydroxylase), polypeptide 1; hCYP39A1; Oxysterol 7-alpha-hydroxylase
Gene Aliases: CYP39A1
UniProt ID: (Human) Q9NYL5
Entrez Gene ID: (Human) 51302
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.