Novus Biologicals
Manufacturer Code:NBP250205UV
Catalog # NBP250205UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V32-P5E5 |
Immunogen | Ovalbumin-conjugated synthetic peptide - RYRLLPPPGV (C-terminal sequence) |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CYPIIW1; CYPIIW1 cytochrome P450 2W1 cytochrome P450 family 2 subfamily W polypeptide 1 EC 1.14.14.- EC 1.14.14.1 EC 1.14.14.5 FLJ20359 MGC34287; Cytochrome P450 2W1; cytochrome P450, family 2, subfamily W, polypeptide 1
Gene Aliases: CYP2W1
UniProt ID: (Human) Q8TAV3
Entrez Gene ID: (Human) 54905
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.