Invitrogen
Manufacturer Code:PA5101314
Catalog # PIPA5101314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Cytochrome P450 2S1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200011C15Rik AU041727 C79779 CYP2S1 CYPIIS1 Cytochrome P450 2S1 cytochrome P450 family 2 subfamily S member 1 cytochrome P450 2s1 cytochrome P450 family 2 subfamily s polypeptide 1 cytochrome P540 subfamily IIS polypeptide 1 Hydroperoxy icosatetraenoate dehydratase Thromboxane-A synthase UNQ891/PRO1906; CYPIIS1; Cytochrome P450 2S1; cytochrome P450, 2s1; cytochrome P450, family 2, subfamily S, polypeptide 1; cytochrome P540, subfamily IIS, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; Thromboxane-A synthase
Gene Aliases: 1200011C15Rik; AU041727; C79779; CYP2S1; CYPIIS1; UNQ891/PRO1906
UniProt ID: (Human) Q96SQ9, (Mouse) Q9DBX6
Entrez Gene ID: (Human) 29785, (Mouse) 74134
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.