Novus Biologicals
Manufacturer Code:NBP250204FR
Catalog # NBP250204FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M26-P6H1 |
Immunogen | Ovalbumin-conjugated synthetic peptide QPYLICAERR (C-terminal sequence). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome P450 2R1; Cytochrome P450 2R1 cytochrome P450 family 2 R1 cytochrome P450 family 2 subfamily R polypeptide 1 EC 1.14.13.15 MGC4663 vitamin D 25-hydroxylase; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; Vitamin D 25-hydroxylase
Gene Aliases: CYP2R1
UniProt ID: (Human) Q6VVX0
Entrez Gene ID: (Human) 120227
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.