Invitrogen
Manufacturer Code:PA515061
Catalog # PIPA515061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 464-496 amino acids from the C-terminal region of human CYP2R1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome P450 2R1; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; Vitamin D 25-hydroxylase; Vitamin D 25-hydroxylase Cytochrome P450 2R1 CYP2R1
Gene Aliases: CYP2R1
UniProt ID: (Human) Q6VVX0
Entrez Gene ID: (Human) 120227
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.