Novus Biologicals
Manufacturer Code:NBP292810100UL
Catalog # NB027486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-290 of human CYP2F1 (NP_000765.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2F1 CYP2F CYPIIF1 cytochrome P450 2F1 cytochrome P450 family 2 subfamily F polypeptide 1 cytochrome P450 subfamily IIF polypeptide 1 EC 1.14.14.1 flavoprotein-linked monooxygenase MGC126121 microsomal monooxygenase xenobiotic monooxygenase; CYPIIF1; Cytochrome P450 2F1; cytochrome P450, family 2, subfamily F, polypeptide 1; cytochrome P450, subfamily IIF, polypeptide 1; flavoprotein-linked monooxygenase; microsomal monooxygenase; xenobiotic monooxygenase
Gene Aliases: C2F1; CYP2F; CYP2F1; CYPIIF1
UniProt ID: (Human) P24903
Entrez Gene ID: (Human) 1572
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.