Proteintech
Manufacturer Code:215791AP150UL
Catalog # 50-173-3498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP2F1 Fusion Protein Ag16124 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2F1 CYP2F CYP2F1 CYPIIF1 Cytochrome P450 2F1; CYPIIF1; CYPIIF2; Cytochrome P450 2F1; Cytochrome P450 2F2; cytochrome P450, family 2, subfamily F, polypeptide 1; cytochrome P450, family 2, subfamily f, polypeptide 2; cytochrome P450, subfamily 2F, polypeptide 1; Cytochrome P450, subfamily IIF, polypeptide 1; Cytochrome P450-NAH-2; flavoprotein-linked monooxygenase; microsomal monooxygenase; Naphthalene dehydrogenase; Naphthalene hydroxylase; P450-NAH-2; xenobiotic monooxygenase
Gene Aliases: C2F1; CYP2F; CYP2F1; Cyp2f2; Cyp2f4; CYP4502F4; CYPIIF1
UniProt ID: (Human) P24903, (Rat) O35293
Entrez Gene ID: (Human) 1572, (Rat) 54246
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.