Invitrogen
Manufacturer Code:MA525001
Catalog # PIMA525001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F11 |
Immunogen | Full length human recombit protein of CYP2E1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4-nitrophenol 2-hydroxylase; CYP2E1 cytochrome P450 family 2 subfamily E polypeptide 1 CPE1 CYP2E P450-J P450C2E cytochrome P450 2E1 cytochrome P450 subfamily IIE (ethanol-inducible) polypeptide 1 flavoprotein-linked monooxygenase microsomal monooxygenase xenobiotic monooxygenase; CYPIIE1; Cytochrome P450 2E1; cytochrome P450 subfamily 2e1 (ethanol-inducible); cytochrome P450, 2e1, ethanol inducible; cytochrome P450, family 2, subfamily E, polypeptide 1; cytochrome P450, subfamily 2E, polypeptide 1; cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1; Cytochrome P450-ALC; Cytochrome P450-J; Cytochrome P450RLM6; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450-J; P450RLM6; xenobiotic monooxygenase
Gene Aliases: CPE1; CYP2E; Cyp2e-1; CYP2E1; P450-J; P450C2E
UniProt ID: (Human) P05181, (Mouse) Q05421, (Rat) P05182
Entrez Gene ID: (Human) 1571, (Mouse) 13106, (Rat) 25086
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.