Invitrogen
Manufacturer Code:PA579131
Catalog # PIPA579131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of mouse CYP2E1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-nitrophenol 2-hydroxylase; CYPIIE1; Cytochrome P450 2E1; Cytochrome P450 2E1 4-nitrophenol 2-hydroxylase CYPIIE1 Cytochrome P450-ALC Cytochrome P450-J Cyp2e1 Cyp2e Cyp2e-1; cytochrome P450 subfamily 2e1 (ethanol-inducible); cytochrome P450, 2e1, ethanol inducible; cytochrome P450, subfamily 2E, polypeptide 1; Cytochrome P450-ALC; Cytochrome P450-J; Cytochrome P450RLM6; P450-J; P450RLM6
Gene Aliases: Cyp2e; Cyp2e-1; Cyp2e1
UniProt ID: (Mouse) Q05421, (Rat) P05182
Entrez Gene ID: (Mouse) 13106, (Rat) 25086
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.