Invitrogen
Manufacturer Code:PA529796
Catalog # PIPA529796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 247 and 469 of Human CYP2C8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CPC8 CYPIIC8 MP-12/MP-20 P450 form 1 cytochrome P450 2C8 cytochrome P450 IIC2 cytochrome P450 MP-12 cytochrome P450 MP-20 cytochrome P450 form 1 cytochrome P450 subfamily IIC (mephenytoin 4-hydroxylase) polypeptide 8 flavoprotein-linked monooxygenase microsomal monooxygenase s-; CYPIIC8; Cytochrome P450 2C8; Cytochrome P450 form 1; Cytochrome P450 IIC2; Cytochrome P450 MP-12; Cytochrome P450 MP-20; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450 form 1; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase
Gene Aliases: CPC8; CYP2C8; CYPIIC8; MP-12/MP-20
UniProt ID: (Human) P10632
Entrez Gene ID: (Human) 1558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.