Proteintech
Manufacturer Code:165461AP150UL
Catalog # 50-173-1018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP2C8 Fusion Protein Ag9898 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CPC8 CYP2C8 CYPIIC8 Cytochrome P450 2C8 Cytochrome P450 form 1 Cytochrome P450 IIC2 Cytochrome P450 MP 12 Cytochrome P450 MP 20 MP 12/MP 20 S mephenytoin 4 hydroxylase; CYPIIC8; Cytochrome P450 2C8; Cytochrome P450 form 1; Cytochrome P450 IIC2; Cytochrome P450 MP-12; Cytochrome P450 MP-20; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450 form 1; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase
Gene Aliases: CPC8; CYP2C8; CYPIIC8; MP-12/MP-20
UniProt ID: (Human) P10632
Entrez Gene ID: (Human) 1558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.