OriGene
Manufacturer Code:TA504328
Catalog # 50-167-5045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D5 |
Immunogen | Full length human recombit protein of human CYP2B6 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1,4-cineole 2-exo-monooxygenase; CPB6 CYP2B CYP2B7 CYP2B7P CYPIIB6 EFVM IIB1 P450; CYPIIB11; CYPIIB3; CYPIIB6; Cytochrome P450 2B11; Cytochrome P450 2B3; Cytochrome P450 2B6; Cytochrome P450 IIB1; Cytochrome P450 PBD-2; cytochrome P450, family 2, subfamily B, polypeptide 11; cytochrome P450, family 2, subfamily B, polypeptide 6; cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6; cytochrome P450IIB3; Cytochrome-P450-2B11
Gene Aliases: CPB6; CYP2B; Cyp2b-3; CYP2B11; Cyp2b3; CYP2B6; CYP2B7; CYP2B7P; CYPIIB11; CYPIIB6; EFVM; IIB1; P450
UniProt ID: (Human) P20813, (Dog) P24460, (Rat) P13107
Entrez Gene ID: (Human) 1555, (Dog) 474177, (Rat) 286953
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.