Invitrogen
Manufacturer Code:PA579128
Catalog # PIPA579128
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human CYP27B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1alpha(OH)ase; 25 hydroxyvitamin D3-1-alpha hydroxylase; 25(OH)D 1alpha-hydroxylase; 25-hydroxyvitamin D(3) 1-alpha-hydroxylase; 25-hydroxyvitamin D-1 alpha hydroxylase mitochondrial; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial precursor (25-OHD-1 alpha-hydroxylase) (25-hydroxyvitamin D3 1-alpha-hydroxylase) (VD3 1A hydroxylase) (P450C1 alpha) (P450VD1-alpha); 25-hydroxyvitamin D3 1alpha-hydroxylase; 25-OHD-1 alpha-hydroxylase; Calcidiol 1-monooxygenase; Cytochrome p450 27B1; cytochrome P450 40 (25-hydroxyvitamin D3 1 alpha-hydroxylase); cytochrome P450 subfamily XXVIIB (25-hydroxyvitamin D-1-alpha-hydroxylase) polypeptide 1; Cytochrome P450 subfamily XXVIIB polypeptide 1; cytochrome P450, 27b1; cytochrome P450, 40 (25-hydroxyvitamin D3 1 alpha-hydroxylase); cytochrome P450, family 27, subfamily B, polypeptide 1; cytochrome P450, subfamily 27b, polypeptide 1; Cytochrome P450C1 alpha; Cytochrome P450VD1-alpha; P450C1 alpha; P450VD1-alpha; P450VD1alpha; VD3 1A hydroxylase
Gene Aliases: CP2B; CYP1; CYP1ALPHA; CYP27B; CYP27B1; Cyp40; P450c1; PDDR; VDD1; VDDR; VDDRI; VDR
UniProt ID: (Human) O15528, (Mouse) O35084, (Rat) O35132
Entrez Gene ID: (Human) 1594, (Mouse) 13115, (Rat) 114700
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.