Novus Biologicals
Manufacturer Code:NBP229942
Catalog # NBP229942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This CYP27B1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 482-508 amino acids from the C-terminal region of human CYP27B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1alpha(OH)ase; 1alpha(OH)ase 25 hydroxyvitamin D3-1-alpha hydroxylase 25-hydroxyvitamin D(3) 1-alpha-hydroxylase 25-hydroxyvitamin D-1 alpha hydroxylase mitochondrial 25-OHD-1 alpha-hydroxylase Calcidiol 1-monooxygenase CP2B CYP1 CYP1ALPHA CYP27B Cytochrome p450 27B1 Cytochrome P450 subfamily XXVIIB polypeptide 1 cytochrome P450 family 27 subfamily B polypeptide 1 cytochrome P450 subfamily XXVIIB (25-hydroxyvitamin D-1-alpha-hydroxylase) Cytochrome P450C1 alpha Cytochrome P450VD1-alpha EC 1.14.13.13 P450c1 PDDR polypeptide 1 VD3 1A hydroxylase VDD1 VDDR VDDRI VDR; 25 hydroxyvitamin D3-1-alpha hydroxylase; 25-hydroxyvitamin D(3) 1-alpha-hydroxylase; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial; 25-OHD-1 alpha-hydroxylase; Calcidiol 1-monooxygenase; Cytochrome p450 27B1; Cytochrome P450 subfamily XXVIIB polypeptide 1; cytochrome P450, family 27, subfamily B, polypeptide 1; Cytochrome P450C1 alpha; Cytochrome P450VD1-alpha; VD3 1A hydroxylase
Gene Aliases: CP2B; CYP1; CYP1ALPHA; CYP27B; CYP27B1; P450c1; PDDR; VDD1; VDDR; VDDRI; VDR
UniProt ID: (Human) O15528
Entrez Gene ID: (Human) 1594
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.