Novus Biologicals
Manufacturer Code:NBP276660B
Catalog # NBP276660B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V29-P4B8&asteriskB2 |
Immunogen | Ovalbumin-conjugated synthetic peptide KVVLAPETGEL (C-terminal sequence) |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 12-alpha-triol 27-hydroxylase 7-alpha cholestanetriol 26-monooxygenase CP275-beta-cholestane-3-alpha 7-alpha 12-alpha-triol 27-hydroxylase CTX5-beta-cholestane-3-alpha CYP275-beta-cholestane-3-alpha 7-alpha 12-alpha-triol 26-hydroxylase Cytochrome P450 27 cytochrome P450 family 27 subfamily A polypeptide 1 cytochrome P450 subfamily XXVIIA (steroid 27-hydroxylase cerebrotendinousxanthomatosis) polypeptide 1 Cytochrome P-450C27/25 EC 1.14.13.15 sterol 26-hydroxylase mitochondrial Sterol 27-hydroxylase Vitamin D(3) 25-hydroxylase; 5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase; 5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase; 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 26-hydroxylase; 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase; cholestanetriol 26-monooxygenase; Cytochrome P-450C27/25; Cytochrome P450 27; cytochrome P450, family 27, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinous xanthomatosis), polypeptide 1; Sterol 26-hydroxylase, mitochondrial; Sterol 27-hydroxylase; Vitamin D(3) 25-hydroxylase
Gene Aliases: CP27; CTX; CYP27; CYP27A1
UniProt ID: (Human) Q02318
Entrez Gene ID: (Human) 1593
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.