Invitrogen
Manufacturer Code:PA575859
Catalog # PIPA575859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human CYP26C1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome P450 26C1; Cytochrome P450 26C1 (EC:1.14.-.-) CYP26C1; cytochrome P450, family 26, subfamily C, polypeptide 1; predicted gene, EG546726
Gene Aliases: CYP26C1; EG546726; FFDD4
UniProt ID: (Human) Q6V0L0
Entrez Gene ID: (Human) 340665, (Rat) 308190, (Mouse) 546726
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.