Novus Biologicals
Manufacturer Code:NBP250193F
Catalog # NBP250193F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F27P6A1 |
Immunogen | Ovalbumin-conjugated synthetic peptide PARFTHFHGE (C-terminal sequence) |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CP26 CYP26Retinoic acid 4-hydroxylase cytochrome P450 26A1 Cytochrome P450 retinoic acid-inactivating 1 cytochrome P450 family 26 subfamily A polypeptide 1 cytochrome P450 subfamily XXVIA polypeptide 1 Cytochrome P450RAI EC 1.14 hP450RAI P450 retinoic acid-inactivating 1 P450RAI P450RAI1Retinoic acid-metabolizing cytochrome; Cytochrome P450 26A1; Cytochrome P450 retinoic acid-inactivating 1; cytochrome P450, family 26, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIA, polypeptide 1; Cytochrome P450RAI; hP450RAI; P450, retinoic acid-inactivating, 1; Retinoic acid 4-hydroxylase; Retinoic acid-metabolizing cytochrome
Gene Aliases: CP26; CYP26; CYP26A1; P450RAI; P450RAI1
UniProt ID: (Human) O43174
Entrez Gene ID: (Human) 1592
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.