Invitrogen
Manufacturer Code:PA521704
Catalog # PIPA521704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 241 and 514 of CYP24A1 (Uniprot IDNo.Q07973) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,25-@dihydroxyvitamin D3 24-hydroxylase; 1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial; 24-OHase; 25-hydroxyvitamin D-24-hydroxylase; CP24 CYP24 HCAI P450-CC24 125-&commatdihydroxyvitamin D3 24-hydroxylase 125-dihydroxyvitamin D(3) 24-hydroxylase mitochondrial 24-OHase cytochrome P450 24A1 cytochrome P450 subfamily XXIV (vitamin D 24-hydroxylase) cytochrome P450-CC24 exo-mitochondrial protein vitamin D 24-hydroxylase vitamin D(3) 24-hydroxylase; Cytochrome P450 24A1; cytochrome P450, 24; cytochrome P450, 24a1; cytochrome P450, family 24, subfamily A, polypeptide 1; cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase); Cytochrome P450-CC24; exo-mitochondrial protein; vitamin D 24-hydroxylase; vitamin D(3) 24-hydroxylase
Gene Aliases: 24-OHase; CP24; Cyp-24; CYP24; CYP24A1; HCAI; HCINF1; P450-CC24
UniProt ID: (Human) Q07973, (Mouse) Q64441
Entrez Gene ID: (Human) 1591, (Mouse) 13081
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.