Invitrogen
Manufacturer Code:PA519406
Catalog # PIPA519406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RKYDIQATDN) corresponding to the internal amino acids of CYP24A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,25-@dihydroxyvitamin D3 24-hydroxylase; 1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial; 24-OHase; 24R-Ohase; 25-hydroxyvitamin D3 24-hydroxylase; CYP24A1 cytochrome P450 family 24 subfamily A polypeptide 1 CP24 CYP24 MGC126273 MGC126274 P450-CC24 125-&commatdihydroxyvitamin D3 24-hydroxylase 24-OHase cytochrome P450 family 24 subfamily A polypeptide 1 cytochrome P450 subfamily XXIV (vitamin D 24-hydroxylase) exo-mitochondrial protein vitamin D 24-hydroxylase; Cytochrom P450 (25-hydroxyvitamin D24-hydroxylase); Cytochrome P450 24A1; cytochrome P450, family 24, subfamily A, polypeptide 1; cytochrome P450, subfamily 24; cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase); Cytochrome P450-CC24; exo-mitochondrial protein; P450-CC24; vitamin D 24-hydroxylase; vitamin D(3) 24-hydroxylase
Gene Aliases: 25-hydroxyvitaminD324-hydroxylase; CP24; CYP24; CYP24A1; HCAI; HCINF1; P450-CC24; VITD12
UniProt ID: (Human) Q07973, (Rat) Q09128
Entrez Gene ID: (Human) 1591, (Rat) 25279
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.