Proteintech
Manufacturer Code:167021AP150UL
Catalog # 50-173-1126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CYP20A1 Fusion Protein Ag10099 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYP M CYP20A1 Cytochrome P450 20A1; Cytochrome P450 20A1; cytochrome P450 monooxygenase; cytochrome P450, 20a1; cytochrome P450, family 20, subfamily A, polypeptide 1
Gene Aliases: A930011N14Rik; CYP-M; CYP20A1; Cypm; UNQ667/PRO1301
UniProt ID: (Human) Q6UW02, (Mouse) Q8BKE6, (Rat) Q6P7D4
Entrez Gene ID: (Human) 57404, (Mouse) 77951, (Rat) 316435
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.