Invitrogen
Manufacturer Code:PA528040
Catalog # PIPA528040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 224 and 492 of Human Cytochrome P450 1B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aryl hydrocarbon hydroxylase; benz[a]anthracene inducible; CP1B CYPIB1 GLC3A P4501B1 aryl hydrocarbon hydroxylase cytochrome P450 1B1 cytochrome P450 subfamily I (dioxin-inducible) polypeptide 1 (glaucoma 3 primary infantile) flavoprotein-linked monooxygenase microsomal monooxygenase xenobiotic monooxygenase; CYPIB1; Cytochrome P450 1B1; cytochrome P450, 1b1, benz[a]anthracene inducible; cytochrome P450, family 1, subfamily B, polypeptide 1; cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, primary infantile); Cytochrome P450CMEF; Cytochrome P450EF; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; xenobiotic monooxygenase
Gene Aliases: CP1B; Cyp1-b1; CYP1B1; CYPIB1; GLC3A; P4501B1
UniProt ID: (Human) Q16678, (Mouse) Q64429
Entrez Gene ID: (Human) 1545, (Mouse) 13078
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.