Invitrogen
Manufacturer Code:PA515058
Catalog # PIPA515058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 143-176 amino acids from the central region of human CYP1B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aryl hydrocarbon hydroxylase; CYPIB1; Cytochrome P450 1B1; Cytochrome P450 1B1 CYPIB1 CYP1B1; cytochrome P450, family 1, subfamily B, polypeptide 1; cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, primary infantile); flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; xenobiotic monooxygenase
Gene Aliases: CP1B; CYP1B1; CYPIB1; GLC3A; P4501B1
UniProt ID: (Human) Q16678
Entrez Gene ID: (Human) 1545
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.