Novus Biologicals
Manufacturer Code:NBP297885C
Catalog # NB048951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human CYP1B1 fragment. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aryl hydrocarbon hydroxylase; aryl hydrocarbon hydroxylase CP1B CYPIB1 cytochrome P450 1B1 cytochrome P450 family 1 subfamily B polypeptide 1 cytochrome P450 subfamily I (dioxin-inducible) polypeptide 1 (glaucoma 3 EC 1.14.14.1 flavoprotein-linked monooxygenase GLC3A microsomal monooxygenase P4501B1 primary infantile) xenobiotic monooxygenase; CYPIB1; Cytochrome P450 1B1; cytochrome P450, family 1, subfamily B, polypeptide 1; cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, primary infantile); flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; xenobiotic monooxygenase
Gene Aliases: CP1B; CYP1B1; CYPIB1; GLC3A; P4501B1
UniProt ID: (Human) Q16678
Entrez Gene ID: (Human) 1545
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.